COMH4211AF70 Combi-Blocks, Inc.

3-[(2-Chloro-4-fluorobenzyl)oxy]azetidine

  • MDL number MFCD11874027
  • Cas Number : 1121598-13-7
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2930
Availability details for
Quantity
  • Availability
  • Add to Cart