COMH4211AF71 Combi-Blocks, Inc.

(R)-1-(2-Fluoro-6-methoxy-benzyl)-piperidin-3-ylamine hydrochloride

  • MDL number MFCD31613750
  • Cas Number : 2270918-24-4
  • Molecular Weight : 274.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2933
Availability details for
Quantity
  • Availability
  • Add to Cart