COMH4211AF72 Combi-Blocks, Inc.

Bupropion 3',4'-dichloro impurity

  • MDL number MFCD22683736
  • Cas Number : 1193779-34-8
  • Molecular Weight : 310.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2934
Availability details for
Quantity
  • Availability
  • Add to Cart