COMH4211AF77 Combi-Blocks, Inc.

1-Cyclopropylmethyl-5-fluoro-1h-indole-2-carboxylic acid

  • MDL number MFCD31579377
  • Cas Number : 1781345-86-5
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2947
Availability details for
Quantity
  • Availability
  • Add to Cart