COMH4211AF7A Combi-Blocks, Inc.

1-[(2-Chloro-5-nitrophenyl)methyl]-4-methylpiperazine

  • MDL number MFCD01466580
  • Cas Number : 414880-12-9
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2955
Availability details for
Quantity
  • Availability
  • Add to Cart