COMH4211AF7C Combi-Blocks, Inc.

(2-Amino-5-bromophenyl)[(1r)-1-phenylethyl]amine

  • MDL number MFCD30729449
  • Cas Number : 1416336-80-5
  • Molecular Weight : 291.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2958
Availability details for
Quantity
  • Availability
  • Add to Cart