COMH4211AF7F Combi-Blocks, Inc.

4-(3-Bromophenoxy)-1-butanol

  • MDL number MFCD12186908
  • Cas Number : 157245-85-7
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2964
Availability details for
Quantity
  • Availability
  • Add to Cart