COMH4211AF80 Combi-Blocks, Inc.

4-(2,2-Dimethylpropanoyl)benzoic acid

  • MDL number MFCD09965971
  • Cas Number : 32018-31-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2965
Availability details for
Quantity
  • Availability
  • Add to Cart