COMH4211AF82 Combi-Blocks, Inc.

6-Chloro-5-methoxypicolinaldehyde

  • MDL number MFCD16606538
  • Cas Number : 1227514-34-2
  • Molecular Weight : 171.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2967
Availability details for
Quantity
  • Availability
  • Add to Cart