COMH4211AF83 Combi-Blocks, Inc.

(4-Amino-3-fluoro-phenoxy)-acetic acid tert-butyl ester

  • MDL number MFCD31579285
  • Cas Number : 1162676-58-5
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2968
Availability details for
Quantity
  • Availability
  • Add to Cart