COMH4211AF84 Combi-Blocks, Inc.

1-[5-Fluoro-2-(2-methoxy-ethoxy)-phenyl]-ethylamine

  • MDL number MFCD11137272
  • Cas Number : 1019573-25-1
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2971
Availability details for
Quantity
  • Availability
  • Add to Cart