COMH4211AF88 Combi-Blocks, Inc.

4-Chloro-2-fluoro-6-methylaniline

  • MDL number MFCD16817663
  • Cas Number : 939989-99-8
  • Molecular Weight : 159.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2977

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2977
Availability details for
Quantity
  • Availability
  • Add to Cart