COMH4211AF8A Combi-Blocks, Inc.

5-Chloro-2-(4-iodophenoxy)pyrimidine

  • MDL number MFCD25575705
  • Cas Number : 1519620-21-3
  • Molecular Weight : 332.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-(4-iodophenoxy)pyrimidine
Inchi Code 1S/C10H6ClIN2O/c11-7-5-13-10(14-6-7)15-9-3-1-8(12)2-4-9/h1-6H
Inchi Key QEOLZFZKKCLKHS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2983

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2983
Availability details for
Quantity
  • Availability
  • Add to Cart