COMH4211AF8C Combi-Blocks, Inc.

3-Ethoxy-2-methyl-propan-1-ol

  • MDL number MFCD26084764
  • Cas Number : 53892-30-1
  • Molecular Weight : 118.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2987
Availability details for
Quantity
  • Availability
  • Add to Cart