COMH4211AF8F Combi-Blocks, Inc.

tert-Butyl 4-(3-fluoro-4-(methoxycarbonyl)phenyl)piperazine-1-carboxylate

  • MDL number MFCD11872653
  • Cas Number : 1121599-68-5
  • Molecular Weight : 338.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2993
Availability details for
Quantity
  • Availability
  • Add to Cart