COMH4211AF91 Combi-Blocks, Inc.

1-Bromo-4-fluoro-2-(2,2,2-trifluoroethoxy)benzene

  • MDL number MFCD24122574
  • Cas Number : 870063-18-6
  • Molecular Weight : 273.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2996
Availability details for
Quantity
  • Availability
  • Add to Cart