COMH4211AF92 Combi-Blocks, Inc.

4-[(3-Aminopropyl)sulfanyl]-1,2-dichlorobenzene

  • MDL number MFCD11585823
  • Cas Number : 119511-46-5
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2997
Availability details for
Quantity
  • Availability
  • Add to Cart