COMH4211AF93 Combi-Blocks, Inc.

1-[2-Bromo-5-(trifluoromethyl)phenyl]ethan-1-amine oxalate

  • MDL number MFCD30529960
  • Cas Number : 2205384-01-4
  • Molecular Weight : 358.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3001
Availability details for
Quantity
  • Availability
  • Add to Cart