COMH4211AF94 Combi-Blocks, Inc.

(1H-Pyrazol-4-yloxy)-acetic acid ethyl ester

  • MDL number MFCD31579326
  • Cas Number : 2118649-93-5
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3002
Availability details for
Quantity
  • Availability
  • Add to Cart