COMH4211AF97 Combi-Blocks, Inc.

2-(4-Isobutoxy-pyrazol-1-yl)-2-methyl-propionic acid

  • MDL number MFCD31579342
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3008
Availability details for
Quantity
  • Availability
  • Add to Cart