COMH4211AF9B Combi-Blocks, Inc.

4-(5-Amino-3-tert-butyl-1h-pyrazol-1-yl)benzonitrile

  • MDL number MFCD11869248
  • Cas Number : 869663-55-8
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3021
Availability details for
Quantity
  • Availability
  • Add to Cart