COMH4211AF9E Combi-Blocks, Inc.

[4-[(2-Cyanobenzyl)thio]phenyl]boronic acid

  • MDL number MFCD31614117
  • Cas Number : 1005206-26-7
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3028
Availability details for
Quantity
  • Availability
  • Add to Cart