COMH4211AF9F Combi-Blocks, Inc.

(S)-2-Benzyloxycarbonylamino-3-thiophen-2-yl-propionic acid 2,5-dioxo-pyrrolidin-1-yl ester

  • MDL number MFCD31381635
  • Cas Number : 1799661-04-3
  • Molecular Weight : 402.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3029
Availability details for
Quantity
  • Availability
  • Add to Cart