COMH4211AFA1 Combi-Blocks, Inc.

Trans isopropyl 4-aminocyclohexanecarboxylate hydrochloride

  • MDL number MFCD31583364
  • Cas Number : 2270913-10-3
  • Molecular Weight : 221.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3032
Availability details for
Quantity
  • Availability
  • Add to Cart