COMH4211AFA3 Combi-Blocks, Inc.

N-(2-Aminoethyl)-4-hydroxy-2-methylbenzamide

  • MDL number MFCD31613837
  • Cas Number : 1697749-95-3
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3036
Availability details for
Quantity
  • Availability
  • Add to Cart