COMH4211AFA5 Combi-Blocks, Inc.

1,4-Dibromo-2-fluoro-6-nitrobenzene

  • MDL number MFCD28784828
  • Cas Number : 1807182-22-4
  • Molecular Weight : 298.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3038
Availability details for
Quantity
  • Availability
  • Add to Cart