COMH4211AFA9 Combi-Blocks, Inc.

2,2,2-Trifluoro-1-(piperidin-4-yl)ethan-1-ol hydrochloride

  • MDL number MFCD18800779
  • Cas Number : 184042-84-0
  • Molecular Weight : 219.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3055
Availability details for
Quantity
  • Availability
  • Add to Cart