COMH4211AFAB Combi-Blocks, Inc.

N-Methyl-3-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-ylmethyl]-benzamide

  • MDL number MFCD31583135
  • Molecular Weight : 341.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3058
Availability details for
Quantity
  • Availability
  • Add to Cart