COMH4211AFAC Combi-Blocks, Inc.

2-Thiophenecarboxaldehyde, 4-(4-methylphenyl)-

  • MDL number MFCD06802073
  • Cas Number : 853311-16-7
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3063
Availability details for
Quantity
  • Availability
  • Add to Cart