COMH4211AFAE Combi-Blocks, Inc.

4-Bromo-3-methyl-n-(methylsulfonyl)benzamide

  • MDL number MFCD31557924
  • Cas Number : 166386-80-7
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3065
Availability details for
Quantity
  • Availability
  • Add to Cart