COMH4211AFB1 Combi-Blocks, Inc.

4-Ethoxy-3-methylbenzaldehyde

  • MDL number MFCD06247361
  • Cas Number : 56917-14-7
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3075
Availability details for
Quantity
  • Availability
  • Add to Cart