COMH4211AFB3 Combi-Blocks, Inc.

tert-Butyl n-[2-(4-formylphenoxy)ethyl]carbamate

  • MDL number MFCD11926521
  • Cas Number : 198990-09-9
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3078
Availability details for
Quantity
  • Availability
  • Add to Cart