COMH4211AFB4 Combi-Blocks, Inc.

5-(Cyclopropylmethoxy)-2-isopropoxybenzoic acid

  • MDL number MFCD29099903
  • Cas Number : 927908-18-7
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3080
Availability details for
Quantity
  • Availability
  • Add to Cart