COMH4211AFB5 Combi-Blocks, Inc.

3-tert-Butyl-4-methoxymethoxy-phenol

  • MDL number MFCD31613933
  • Cas Number : 2270911-97-0
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3082
Availability details for
Quantity
  • Availability
  • Add to Cart