COMH4211AFB6 Combi-Blocks, Inc.

2-Acetylsulfanyl-2-methylpropionic acid ethyl ester

  • MDL number MFCD28362805
  • Cas Number : 1151713-51-7
  • Molecular Weight : 190.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3083
Availability details for
Quantity
  • Availability
  • Add to Cart