COMH4211AFB8 Combi-Blocks, Inc.

2-(3-tert-Butyl-4-methoxymethoxy-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

  • MDL number MFCD28166849
  • Cas Number : 1442430-86-5
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3087
Availability details for
Quantity
  • Availability
  • Add to Cart