COMH4211AFB9 Combi-Blocks, Inc.

1-(2-(tert-Butoxy)-2-oxoethyl)-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD28532898
  • Cas Number : 1780873-47-3
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3090
Availability details for
Quantity
  • Availability
  • Add to Cart