COMH4211AFBB Combi-Blocks, Inc.

1-(Tetrahydro-pyran-4-yl)-1h-pyrazol-4-ol

  • MDL number MFCD26129287
  • Cas Number : 1598643-02-7
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-ol
Inchi Code 1S/C8H12N2O2/c11-8-5-9-10(6-8)7-1-3-12-4-2-7/h5-7,11H,1-4H2
Inchi Key OZKMPCJLNKXVGB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3097
Availability details for
Quantity
  • Availability
  • Add to Cart