COMH4211AFBC Combi-Blocks, Inc.

tert-Butyl (2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl)ethyl)carbamate

  • MDL number MFCD28717087
  • Cas Number : 1414475-01-6
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3099
Availability details for
Quantity
  • Availability
  • Add to Cart