COMH4211AFBD Combi-Blocks, Inc.

2-Fluoro-4'-hydroxy-[1,1'-biphenyl]-3-carbaldehyde

  • MDL number MFCD31583134
  • Cas Number : 1611443-83-4
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3102
Availability details for
Quantity
  • Availability
  • Add to Cart