COMH4211AFBF Combi-Blocks, Inc.

Cyclopropyl-(4-isobutoxy-benzyl)-amine

  • MDL number MFCD11629936
  • Cas Number : 1095127-65-3
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3113
Availability details for
Quantity
  • Availability
  • Add to Cart