COMH4211AFC0 Combi-Blocks, Inc.

5-Bromo-2-propoxybenzamide

  • MDL number MFCD07551178
  • Cas Number : 882093-01-8
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3115
Availability details for
Quantity
  • Availability
  • Add to Cart