COMH4211AFC1 Combi-Blocks, Inc.

(2R)-2-(([(4-Chlorophenyl)amino]carbonyl)amino)propanoic acid

  • MDL number MFCD08448704
  • Cas Number : 1396994-81-2
  • Molecular Weight : 242.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3116
Availability details for
Quantity
  • Availability
  • Add to Cart