COMH4211AFC3 Combi-Blocks, Inc.

4-Methanesulfonyl-piperazine-1,2-dicarboxylic acid 1-tert-butyl ester

  • MDL number MFCD31579557
  • Cas Number : 939997-64-5
  • Molecular Weight : 308.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3124
Availability details for
Quantity
  • Availability
  • Add to Cart