COMH4211AFC4 Combi-Blocks, Inc.

5-(4,4-Difluoro-piperidin-1-yl)-pentanoic acid hydrochloride

  • MDL number MFCD31579386
  • Cas Number : 2270905-50-3
  • Molecular Weight : 257.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4,4-difluoropiperidin-1-yl)pentanoic acid hydrochloride
Inchi Code 1S/C10H17F2NO2.ClH/c11-10(12)4-7-13(8-5-10)6-2-1-3-9(14)15;/h1-8H2,(H,14,15);1H
Inchi Key PARFGGAHDZQNEZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3126
Availability details for
Quantity
  • Availability
  • Add to Cart