COMH4211AFC6 Combi-Blocks, Inc.

1-[2-Fluoro-4-(trifluoromethyl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD30725581
  • Cas Number : 1373866-25-1
  • Molecular Weight : 243.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3129
Availability details for
Quantity
  • Availability
  • Add to Cart