COMH4211AFC8 Combi-Blocks, Inc.

2-Bromo-3-(2-methoxy-ethoxy)-benzaldehyde

  • MDL number MFCD31613854
  • Cas Number : 1995743-35-5
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3136
Availability details for
Quantity
  • Availability
  • Add to Cart