COMH4211AFC9 Combi-Blocks, Inc.

2-(Difluoromethoxy)-6-nitrotoluene

  • MDL number MFCD18395256
  • Cas Number : 1261440-78-1
  • Molecular Weight : 203.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3140
Availability details for
Quantity
  • Availability
  • Add to Cart