COMH4211AFCA Combi-Blocks, Inc.

1-(3-Bromo-2-methoxyphenyl)ethanamine hydrochloride

  • MDL number MFCD31539528
  • Cas Number : 2203140-38-7
  • Molecular Weight : 266.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3141
Availability details for
Quantity
  • Availability
  • Add to Cart