COMH4211AFCB Combi-Blocks, Inc.

(R)-2-Amino-n-methyl-4-methylpentanamide

  • MDL number MFCD19160599
  • Cas Number : 312968-66-4
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3143
Availability details for
Quantity
  • Availability
  • Add to Cart